Learn More
CD68 Mouse anti-Human, Unlabeled, Clone: Y1/82A, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 556059
Description
The Y1/82A monoclonal antibody specifically binds to a 110 kDa-type I-transmembrane glycoprotein expressed in cytoplasmic granules of monocytes, macrophages, dendritic cells, granulocytes, myeloid progenitor cells and, reportedly, a subset of CD34-positive hemopoietic bone marrow progenitor cells. CD68 belongs to the sialomucin family and its function has not been fully elucidated. This antibody could be useful in studies of myeloid cell development.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD68 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| PHA-stimulated Human PBMC | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| Y1/82A | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only.