Learn More
CD64 Mouse anti-Human, Alexa Fluor 700, Clone: 10.1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561188
Description
The 10.1 monoclonal antibody specifically binds to CD64, a 72 kDa type I transmembrane glycoprotein that is a high affinity receptor for human IgG (FcγRI), especially the IgG1 and IgG3 subclasses. CD64 is expressed on monocytes, macrophages, dendritic cells, granulocytes activated with interferon-gamma and early myeloid lineage cells. CD64 associates with a signaling FcRγ homodimer to form the functional high affinity FcγRI complex. CD64 functions in both innate and adaptive immune responses and mediates endocytosis, phagocytosis, antigen presentation, antibody-dependent cellular toxicity, cytokine release and superoxide generation.
Specifications
| CD64 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
| Human Rheumatoid synovial fluid cells and fibronectin-purified monocytes | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 10.1 | |
| FCGR1; FcRI; Fc-gamma RI; IgG Fc Receptor I; High affinity IgG FcR1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.