Learn More
CD63 Mouse anti-Human, PE, Clone: H5C6, BD
Description
The H5C6 monoclonal antibody reacts with CD63. CD63 is a 53 kDa, type III lysosomal glycoprotein, expressed on activated platelets, monocytes and macrophages. This molecule is also referred to in the literature as LIMP, gp55, melanoma-associated antigen ME491, Pltgp40, LAMP-3 and is a member of the tetraspan transmembrane 4 superfamily (TM4SF). It is widely expressed on surface and in the cytoplasm of various hematopoietic (monocytes, macrophages) and non-hematopoietic cells (endothelium, fibroblasts, osteoclasts, smooth muscle).
Clone H5C6 also crossreacts with thrombin-activated peripheral blood platelets of baboon, and both rhesus and cynomolgus macaque monkeys. The distribution of H5C6+ activated platelets is similar to that observed with activated platelets from peripheral blood of normal human donors.
Specifications
Specifications
| Antigen | CD63 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | H5C6 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human Splenic Adherent Cells |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.