Learn More
CD62P Mouse anti-Human, Brilliant Violet 421, Clone: AK-4, BD
Mouse Monoclonal Antibody
£152.54 - £361.57
Specifications
| Antigen | CD62P |
|---|---|
| Clone | AK-4 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Violet 421 |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15803309
|
BD Biosciences
564037 |
25 Tests |
£156.00 £152.54 / Each Save £3.46 2% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15813309
|
BD Biosciences
564038 |
100 Tests |
£366.00 £361.57 / Each Save £4.43 1% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.
The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue™ conjugates.
Specifikationer
| CD62P | |
| Flow Cytometry | |
| Brilliant Violet 421 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| AK-4 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| P-Selectin; GMP-140; PADGEM |
For Research Use Only.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.