Learn More
CD62L (L-selectin) Mouse anti-Human, FITC, Clone: SK11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 347443
Description
CD62L, clone SK11, is derived from hybridization of mouse NS-1 myeloma cells with spleen cells from BALB/c mice immunized with peripheral blood T lymphocytes.
The CD62L antigen, Mr 80 kilodaltons (kd), is the leucocyte endothelial cellular adhesion molecule (LECAM). The CD62L antigen belongs to the selectin family of cell adhesion molecules. The CD62L molecule is the human homologue of the murine lymph node homing receptor, MEL 14.
Specifications
| CD62L (L-selectin) | |
| Monoclonal | |
| 12.5μg/mL | |
| Phosphate buffered saline with gelatin and 0.1% sodium azide. | |
| Human Peripheral Blood T Lymphocytes | |
| RUO | |
| Primary | |
| Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
| Flow Cytometry | |
| SK11 | |
| FITC | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.