Learn More
CD62E Rat anti-Mouse, PE, Clone: 10E9.6, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553751
Description
The 10E9.6 monoclonal antibody specifically binds to the 97-110 kDa cell surface glycoprotein E-selectin (CD62E), also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1), which is expressed on endotoxin- or cytokine-stimulated mouse endothelial cells. A suspension of TNF α stimulated mouse brain capillary endothelioma cells, from the cell line bEnd.3, was used as the immunogen. The epitope recognized by mAb 10E9.6 has been mapped to the first and/or second complement regulatory protein repeat domains of E-selectin. The 10E9.6 antibody has been reported to block binding of a monocyte cell line to E-selectin in vitro and to block neutrophil migration in BALB/c, but not C57BL/6 mice. It has no effect on leukocyte rolling in TNF α -treated mouse venules or on in vitro adhesion of myeloid cells to E-selectin. Studies have demonstrated that Cutaneous Lymphocyte Antigen (CLA), recognized by mAb HECA-452 (Cat. no. 555946), may be a ligand for CD62E.
Specifications
| CD62E | |
| Monoclonal | |
| 0.2mg/mL | |
| Sele; E-selectin; Elam; ELAM-1; LECAM2; LYAM2 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 10E9.6 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse brain capillary endothelioma bEnd.3 (TNFα-stimulated) Cell Line | |
| 0.2 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.