Learn More
CD61 Mouse anti-Rat, Unlabeled, Clone: F11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554951
Description
The F11 antibody reacts with the integrin β3 chain (CD61), which associates with the integrin αv chain (CD51), to form the vitronectin receptor found on endothelial cells, myeloid cells, and osteoclasts, and with the αIIb chain (CD41) on platelets and megakaryocytes. Both receptors mediate adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, and von Willebrand factor. F11 mAb has been reported to block the in vitro attachment of osteoclasts to several ligands. Weak reactivity with human osteoclasts, megakaryocytes, and platelets has also been observed. Other reported applications include immunoprecipitation, in vitro blocking, and immunohistochemical staining (IHC) of acetone-fixed frozen and zinc-fixed paraffin-embedded sections. IHC of formalin-fixed paraffin-embedded sections is not recommended.
Specifications
| CD61 | |
| Monoclonal | |
| 0.5mg/mL | |
| Integrin β3 chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| F11 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat bone cell suspensions | |
| 0.5 mg | |
| Immunology | |
| Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.