Learn More
CD57 Mouse anti-Human, FITC, Clone: TB01, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA181071
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD57 antibody, clone TB01 recognizes CD57, also known as HNK-1, an oligosaccharide antigenic determit present on a variety of polypeptides, lipids and chondroitin sulphate proteoglycans.
The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized.
Specifications
| CD57 | |
| Monoclonal | |
| 0.1 mg/mL | |
| B3GAT1 | |
| GlcAT-P, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1 | |
| Mouse | |
| Ion-exchange chromatography | |
| RUO | |
| 27087 | |
| 4°C or -20°C if preferred, store in dark | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| TB01 | |
| FITC | |
| Q9P2W7 | |
| B3GAT1 | |
| Human neuroblastoma cells. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgM |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.