Learn More
CD57 Mouse anti-Human, PE-CF594, Clone: NK-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562488
Description
The NK-1 monoclonal antibody specifically reacts with a 110 kDa carbohydrate antigen associated with myelin-associated glycoprotein expressed on 7-35% of normal peripheral blood lymphocytes including a subset of natural killer cells, a subset of CD8-positive peripheral blood T cells, and on some neural tissues. CD57 is not expressed on granulocytes, platelets, red blood cells or thymocytes. The function of CD57 is still unclear, however, its expression on T-cell subets occurs in late immune responses.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).
Specifications
| CD57 | |
| Monoclonal | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| NK-1 | |
| B3GAT1; Beta-1,3-glucuronyltransferase 1; HNK1; LEU7; NK-1; GLCATP; GLCUATP | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.