Learn More
CD57 Mouse anti-Human, Unlabeled, Clone: HNK-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559048
Description
Anti-HNK-1 monoclonal antibody recognizes a 110 kD glycoprotein (CD57) that is expressed on a subset of natural killer (NK) lymphocytes and T lymphocytes. It also recognizes epitopes on neural or neuroectodermal tumors including neurofibromas, malignant melanomas, malignant peripheral neuroectodermal tumors, Ewing's sarcomas, and small-cell lung carcinomas. Anti-HNK-1 monoclonal antibody also recognizes a carbohydrate epitope on subpopulations of several cell-adhesion molecules including N-CAM and myelin-associated glycoprotein (MAG).
Specifications
| CD57 | |
| Monoclonal | |
| 1.0mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Membrane extracts of the HSB-2 T-lymphoblastoid cell line | |
| 0.2 mg | |
| Immunology | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| HNK-1 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.