Learn More
CD57 Mouse anti-Human, APC, Clone: NK-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560845
Description
The NK-1 monoclonal antibody specifically reacts with a 110 kDa carbohydrate antigen associated with myelin-associated glycoprotein expressed on 7-35% of normal peripheral blood lymphocytes including a subset of natural killer cells, a subset of CD8-positive peripheral blood T cells, and on some neural tissues. CD57 is not expressed on granulocytes, platelets, red blood cells or thymocytes. The function of CD57 is still unclear, however, its expression on T-cell subets occurs in late immune responses.
Specifications
| CD57 | |
| Monoclonal | |
| APC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| NK-1 | |
| B3GAT1; Beta-1,3-glucuronyltransferase 1; HNK1; LEU7; NK-1; GLCATP; GLCUATP | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.