Learn More
CD56 Mouse anti-Human, BUV737, Clone: NCAM16.2 (also known as NCAM 16), BD
Mouse Monoclonal Antibody
£252.00 - £515.00
Specifications
| Antigen | CD56 |
|---|---|
| Clone | NCAM16.2 (also known as NCAM 16) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Ultraviolet 737 |
Description
The NCAM16.2 monoclonal antibody specifically binds to human CD56. It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39. The CD56 antigen is expressed on approximately 10% to 25% of peripheral blood lymphocytes. It is present on essentially all resting and activated CD16+ natural killer (NK) lymphocytes and approximately 5% of CD3+ peripheral blood lymphocytes. CD3+ CD56+ T lymphocytes comprise a unique subset of cytotoxic T lymphocytes that mediates non-major histocompatibility complex (MHC)-restricted cytotoxicity. CD56 antigen density on NK lymphocytes increases upon cellular activation. The CD56 antigen is involved in neuronal homotypic cell adhesion and cell differentiation during embryogenesis. CD16+ CD56+ NK cells demonstrate reciprocal transfer of an activation state with dendritic cells.
The antibody was conjugated to BD Horizon™ BUV737 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye is a tandem fluorochrome of BD Horizon BUV395 with an Ex Max of 348-nm and an acceptor dye with an Em Max at 737-nm. BD Horizon Brilliant BUV737 can be excited by the ultraviolet laser (355 nm) and detected with a 740/35 filter. Due to the excitation of the acceptor dye by other laser lines, there may be significant spillover into channels detecting Alexa Fluor™ 700-like dyes (e.g., 712/20-nm filter).
Specifications
| CD56 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 737 | |
| RUO | |
| Immunoaffinity-enriched adult human brain NCAM | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| NCAM16.2 (also known as NCAM 16) | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG2b κ | |
| Affinity Purified | |
| NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.