Learn More
CD56 Mouse anti-Human, BB515, Clone: B159, BD
Mouse Monoclonal Antibody
£116.76 - £326.74
Specifications
| Antigen | CD56 |
|---|---|
| Clone | B159 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | Brilliant Blue 515 (BB515) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15853679
|
BD Biosciences
564489 |
25 Tests |
£122.00 £116.76 / Each Save £5.24 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15843679
|
BD Biosciences
564488 |
100 Tests |
£341.00 £326.74 / Each Save £14.26 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The B159 monoclonal antibody specifically binds to CD56. CD56 is a heavily glycosylated adhesion protein that is present on a subpopulation of peripheral blood large granular lymphocytes that demonstrate natural killer activity. CD56 is also expressed on a subset of T cells but is not expressed on myeloid cells, erythrocytes or B cells. This antigen is a pan-NK-cell marker. CD56 is virtually identical to an isoform of the neural cell adhesion molecule (NCAM), a structure mediating homotypic and heterotypic cell-cell interactions.
The antibody was conjugated to BD Horizon™ BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g., 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Specifications
| CD56 | |
| Flow Cytometry | |
| Brilliant Blue 515 (BB515) | |
| RUO | |
| Human NK Cells | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| B159 | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity Purified | |
| NCAM1; NCAM-1; Neural cell adhesion molecule 1; NCAM; NKH1; MSK39 |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.