missing translation for 'onlineSavingsMsg'
Learn More

CD55 Hamster anti-Mouse, PE, Clone: RIKO-5, BD

Hamster Monoclonal Antibody

Brand:  BD Biosciences 558037

Product Code. 15889168

  • ONLINE EXCLUSIVE PRICE
    £379.00
    £363.83 / Each
    Save £15.17
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The RIKO-5 antibody reacts with the membrane-associated complement regulator CD55, also known as decayaccelerating factor (DAF).CD55 is expressed on the surface of erythrocytes and most splenic leukocytes, including B and T lymphocytes and CD11b-positive splenocytes,but it is not detected on thymocytes.Immunohistochemical staining (using a different monoclonal antibody) demonstrates that CD55 is expressed on vascular endothelium and many other cells types in a wide variety of organs.In the mouse, there are two isoforms of CD55, transmembrane and GPI-anchored, that are recognized by the RIKO-5 antibody.Both isoforms are able to protect cells from complement attack by inhibiting the deposition of C3 on antibody-sensitized cells,but the RIKO-5 antibody is not able to neutralize this activity. CD97, a member of the EGF-TM7 family that is widely expressed in mouse tissues, is another ligand for CD55.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD55
Monoclonal
0.2mg/mL
DAF, decay-accelerating factor
Hamster
0.1 mg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
RIKO-5
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Mouse
IgG3 λ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.