Learn More
CD51 Rat anti-Mouse, Biotin, Clone: RMV-7, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 551380
Description
The rat anti-mouse CD51 (clone RMV-7) antibody reacts with the 140 kDa integrin αV chain. Heterodimers of CD51 with several integrin β chains function as receptors for extracellular matrix proteins. CD51/CD61 (αVβ3 integrin, vitronectin receptor) mediates adhesion to fibronectin, fibrinogen, vitronectin, thrombospondin, von Willebrand factor, and CD31 (PECAM-1). It has been reported to be expressed on activated T lymphocytes, polymorphonuclear granulocytes, blastocysts, and osteoclasts. CD51 has reportedly been found to be undetectable on mouse platelets using either antibody clones H9.2B8 or RMV-7. CD51 also forms heterodimers with CD29 (integrin β1), integrins β5, β
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD51 | |
| Monoclonal | |
| 0.5mg/mL | |
| Integrin αV chain | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| RMV-7 | |
| Biotin | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse (BALB/c) IL-2-activated killer (LAK) Cells | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.