Learn More
CD50 (ICAM-3) Mouse anti-Human, FITC, Clone: TÜ41, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555958
Description
The TU41 monoclonal antibody specifically binds to the intracellular adhesion molecule-3 (ICAM-3) which is also known as CD50. ICAM-3 is a 110-120 kDa type I membrane glycoprotein expressed on all leukocytes (platelets and erythrocytes are ICAM-3-negative). ICAM-3 is also expressed on epidermal Langerhans cells and endothelial cells. It predominantly interacts with LFA-1 (CD11a/CD18) without requiring activation as the initial dendritic cell-T lymphocyte interaction. ICAM-3 is an important LFA-1 ligand that functions in the initiation of the immune/inflammatory response and also plays a role in chemotaxis.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD50 | |
| Monoclonal | |
| FITC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| TÜ41 | |
| ICAM-3; ICAM3; Intercellular adhesion molecule 3 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.