Learn More
CD5 Mouse anti-Human, PE-Cy5, Clone: UCHT2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555354
Description
The UCHT2 monoclonal antibody specifically binds to CD5. CD5 is a 67 kDa single-chain, type 1 transmembrane glycoprotein expressed on most thymocytes, the majority of peripheral T lymphocytes and a subpopulation of B cells. CD72 has been shown to be the natural ligand for CD5. CD5+ B cells produce polyreactive antibodies (mostly IgM).
Specifications
| CD5 | |
| Monoclonal | |
| PE-Cyanine5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| UCHT2 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human T cells | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.