missing translation for 'onlineSavingsMsg'
Learn More

CD49E Mouse anti-Human, Unlabeled, Clone: VC5, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555651

Product Code. 15878328

  • ONLINE EXCLUSIVE PRICE
    £121.00
    £115.82 / Each
    Save £5.18
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reacts with the α5 integrin, a dimer of 135/25kDa, that associates with β1 integrin (CD29) to form VLA-5, a well-established fibronectin receptor that is expressed on many cell types, including fibroblasts, endothelial cells, epithelial cells, platelets, peripheral T cells, the myeloid cell line U937, K562 and some melanoma cell lines. This clone does not inhibit α5β1/ligand binding. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD49e
Monoclonal
0.5mg/mL
Integrin α5 chain
Mouse
0.1 mg
Immunology
Human, Pig
IgG1 κ
Flow Cytometry
VC5
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.