Learn More
CD49e Mouse anti-Human, Unlabeled, Clone: IIA1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555615
Description
Reacts with the α5 integrin, a dimer of 135/25kDa, that associates with β1 integrin (CD29) to form VLA-5, a well-established fibronectin receptor that is expressed on many cell types, including fibroblasts, endothelial cells, epithelial cells, platelets, peripheral T cells, the myeloid cell line U937, K562 and some melanoma cell lines. This clone inhibits α5β1/ligand binding, and is useful for studies of the expression and function of this integrin. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD49e | |
| Monoclonal | |
| 0.5mg/mL | |
| Integrin α5 Chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| IIA1 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human α5β1 Protein | |
| 0.1 mg | |
| Immunology | |
| Human, Rabbit | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.