missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49e Rat anti-Mouse, Unlabeled, Clone: 5H10-27, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 553319
This item is not returnable.
View return policy
Description
The 5H10-27 (MFR5) antibody reacts with the α; 5 chain of the integrin α;5 β; 1 fibronectin receptor (CD49e/CD29, VLA-5) on thymocytes, activated T lymphocytes, mast cells, and a variety of mouse cell lines, but not splenocytes. Soluble 5H10-27 (MFR5) antibody has been reported to inhibit VLA-5–mediated functions in vitro. In addition, immobilized mAb 5H10-27 (MFR5) has been demonstrated to costimulate the proliferative response of CD8+ T cells to plate-bound anti-CD3e antibody.
Specifications
| CD49e | |
| Monoclonal | |
| 0.5mg/mL | |
| Integrin α5 chain | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| 5H10-27 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse mast cell line MC/9 | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction