missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD49d Mouse anti-Human, NA/LE, Unlabeled, Clone: 9F10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555501
This item is not returnable.
View return policy
Description
Specifications
| CD49d | |
| Monoclonal | |
| 1.0mg/mL | |
| Integrin α4 chain; Integrin alpha 4; ITGA4; IA4; alpha 4 subunit of VLA-4 | |
| Mouse | |
| 0.5 mg | |
| Immunology | |
| Baboon, Canine, Cynomolgus, Equine, Feline, Human, Rhesus Monkey, Sheep | |
| IgG1 κ |
| Flow Cytometry | |
| 9F10 | |
| Unconjugated | |
| No azide/low endotoxin: Aqueous buffered solution containing no preservative, 0.2μm sterile filtered. Endotoxin level is ≤0.01 EU/μg (≤0.001 ng/μg) of protein as determined by the LAL assay. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction