Learn More
CD49b Hamster anti-Mouse, PE, Clone: HMA2, BD
Hamster Monoclonal Antibody
Brand: BD Biosciences 558759
Description
The HM α 2 antibody reacts with integrin α 2 chain (CD49b), the 150-kDa transmembrane glycoprotein that non-covalently associates with the integrin β 1 subunit (CD29) to form the integrin α 2β 1 complex known as VLA-2. VLA-2, a receptor for collagen and laminin, is expressed on some splenic CD4+ T lymphocytes and NK-T cells, intestinal intraepithelial and lamina propria lymphocytes, splenic NK cells, epithelial cells, and platelets; but it is not on thymocytes or Peyer’s-patch or lymphnode lymphocytes. The expression of VLA-2 is upregulated on lymphocytes in response to mitogens. The HM α 2 antibody has been reported to partially block the interaction of T-cell blasts, but not NK cells, with collagen. Purified HM α 2 mAb blocks the staining of splenic NK cells by the anti-CD49b/Pan-NK Cells mAb DX5 (Cat. No. 553858, for the PE conjugate). Therefore, mAb HM α 2 may be used like the DX5 mAb for identification of NK cells.
Specifications
| CD49b | |
| Monoclonal | |
| 0.2mg/mL | |
| Integrin α2 chain | |
| Hamster | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| HMA2 | |
| PE | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse colon carcinoma cell line Colon26 | |
| 0.1 mg | |
| Immunology | |
| Mouse | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.