Learn More
CD49b Rat anti-Mouse, PE-CF594, Clone: DX5, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 562453
Description
The rat anti-mouse CD49b monoclonal antibody (clone DX5) specifically binds t o the integrin α2 chain (CD49b). C D49b is a 150 kDa transmembrane glycoprotein that non-covalently ass ociates with CD29 (integrin β1) to form the integrin α2β1 complex known as VLA-2. The rat anti-mouse CD49b antibody (clone DX5) has been reported to identify the majority of NK cells and a small T-cell subpopulation in most mouse strains (e.g., A/J, AKR, BALB/c, C3H/HeJ, C57BL/6, C57BL/10, C57BR, C58, CBA/Ca, DBA/1, DBA/2, SJL, SWR, 129/J, but not NOD). The DX5 antibody also recognizes platelets that express high levels of CD49b. Multiparameter flow cytometric analysis has demonstrated that most lymphocytes which express NK-1.1 (NKR-P1B and NKR-P1C), as detectable by mouse anti-mouse NK-1.1 antibody (clone PK136), also express the DX5 antigen. Small DX5+ NK-1.1- and DX5- NK-1.1+ cell subsets are found, especially among the CD3-positive cell population. Some CD49b+ NK cells have been reported to gradually lose reactivity with the rat anti-mouse CD49b antibody (clone DX5) when cultured in the presence of recombinant human IL-2. The resulting DX5-negative cells have weakened cytotoxic activity when compared to the remaining DX5+ cells. This indicates that the DX5 antibody distinguishes functional subsets of NK cells. No activation or blocking activity of the rat anti-mouse antibody (clone DX5) has been observed. Staining of splenic NK cells with this antibody reportedly can be blocked by hamster anti-mouse CD49b antibody (clone HMα2).
Specifications
| CD49b | |
| Monoclonal | |
| 0.2mg/mL | |
| itga2; Integrin alpha-2; DX5; Pan NK cell marker; VLAA2; VLA-2 alpha chain | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgM κ |
| Flow Cytometry | |
| DX5 | |
| PE-CF594 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Mouse (C57BL/6) NK1.1+ cells propagated with rIL-2 | |
| 0.1 mg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.