Learn More
CD49b Mouse anti-Human, Unlabeled, Clone: 2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 611016
Description
Integrins are a family of dimeric proteins that mediate cell-to-cell and extracellular matrix adhesion. They consist of a large α chain that is non-covalently associated with a smaller β chain which is used to define integrin subfamilies. These molecules exhibit a wide range of expression throughout development and adulthood. VLA-2 (very late antigen), a member of the integrin superfamily, was identified on activated T cells, but has since been reported to be on various cell types. VLA-2 is reported to be a heterodimer of integrin α2 (CD49b) and integrin β1 (CD29) subunits. The α2 chain contains a large extracellular domain, a transmembrane domain, and a short cytoplasmic tail. VLA-2 functions as a collagen receptor on platelets and fibroblasts, as well as a collagen and laminin receptor on endothelial and epithelial cells. On activated T cells, VLA-2, like LFA-1, exhibits increased number and affinity of ligand binding. Interactions of these molecules with their extracellular matrix ligands is important for directing effector T cells to their target tissues and to provide co-stimulatory signals. Thus, VLA-2 not only plays an important role in cellular adhesion, but may function in intracellular signal transmission.
Immunofluorescence, Western Blotting
Specifications
| CD49b | |
| Monoclonal | |
| 250μg/mL | |
| Integrin α2; VLA-2α | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 2 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Human VLA-2α aa.42-245 | |
| 50 μg | |
| Immunology | |
| Human | |
| IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.