Learn More
CD48 Mouse anti-Human, Unlabeled, Clone: TÜ145, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555758
Description
The TÜ145 monoclonal antibody specifically binds to CD48. CD48 belongs to the CD2 family within the immunoglobulin superfamily. CD48 is a ∽43 kDa glycosylphosphatidylinositol (GPI)-anchored single chain glycoprotein. CD48 is strongly expressed on lymphocytes, monocytes and macrophages and weakly on granulocytes. The expression of CD48 is upregulated following lymphocyte activation and by Epstein-Barr virus (EBV) infection. CD48 binds with high affinity to CD244 and with low affinity to CD2. These receptor-ligand interactions can augment or inhibit leukocyte responses.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD48 | |
| Monoclonal | |
| 0.5mg/mL | |
| BCM1; BLAST; BLAST1; Hulym3; MEM-102; OX45; SLAMF2; TCT.1 | |
| Mouse | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| TÜ145 | |
| Unconjugated | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.