Learn More
CD48 (BCM-1) Hamster anti-Mouse, PE-Cy7, Clone: HM48-1, BD
Description
The HM48-1 monoclonal antibody specifically binds to CD48 (previously known as BCM1 in mice, Blast-1 in human, and OX-45 in the rat), a GPI-anchored member of the Ig superfamily. It is widely distributed on leukocytes, but not on non-hematopoietic cells, and its ligands include CD2 (LFA-2) and CD244 (2B4 antigen). The HM48-1 mAb blocks binding of soluble CD2 to CD48-bearing cells, blocks the interaction of CD2 and CD244, inhibits spleen cell proliferative responses to mitogens, augments the proliferative response of spleen cells when cross-linked with anti-CD3e mAbs, and inhibits priming of CTL in vitro. In vivo administration of HM48-1 antibody can prolong survival of cardiac allografts, an effect which is greatly enhanced by the addition of anti-CD2 mAb 12-15. This hamster mAb to a mouse leukocyte antigen does not cross-react with rat leukocytes.
Specifications
Specifications
| Antigen | CD48 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HM48-1 |
| Concentration | 0.2mg/mL |
| Conjugate | PE-Cyanine7 |
| Description | BLAST; BLAST-1; BCM1; HM48-1; MEM-102; Sgp-60; SLAMF2 |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Host Species | Hamster |
| Immunogen | Mouse T lymphoma MBL-2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.