Learn More
CD46 Mouse anti-Human, PE, Clone: E4.3, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564252
Description
The E4.3 monoclonal antibody specifically binds to CD46. CD46 is also known as membrane cofactor protein (MCP). CD46 is a type I membrane glycoprotein composed of two non-disulfide linked α (66 kDa) and β (56 kDa) chains expressed on lymphocytes, monocytes and granulocytes. It is not expressed on erythrocytes or platelets. There are four isoforms of the dimer which function as complement regulatory factors. CD46 promotes the enzymatic degradation of activated C3b and/or C4b deposited on host cells. It also serves as the measles virus receptor.
Specifications
| CD46 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2a κ |
| Flow Cytometry | |
| E4.3 | |
| MCP; TLX | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.