Learn More
CD45 Mouse anti-Rat, Unlabeled, Clone: OX-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550566
Description
The OX-1 antibody reacts with all molecular forms of CD45 (Leukocyte Common Antigen) on all hematopoietic cells except erythrocytes. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the rat are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction.
Specifications
| CD45 | |
| Monoclonal | |
| 250μg/mL | |
| Ptprc; Lca; Leucocyte common antigen; RT7; T200 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| OX-1 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, goat serum, and ≤0.09% sodium azide. | |
| Leukocyte Common Antigen-enriched Glycoprotein Fraction from Wistar Rat Thymocytes | |
| 1 mL | |
| Immunology | |
| Rat | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.