Learn More
CD44 Mouse anti-Human, PE, Clone: 515, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550989
Description
Reacts with CD44, and 85 kDa glycoprotein that is expressed on leukocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is a member of the hyaladherin family of hyaluronan-binding proteins, with structural similarities to selectins. It has a functional role in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. 515 monoclonal antibody has been reportedto be able to block cell adhesion to the hyaluronic acid.
Specifications
| CD44 | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 515 | |
| Pgp-1, H-CAM, Ly24 | |
| Mouse | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.