Learn More
CD44 Mouse anti-Human, Unlabeled, Clone: L178, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 559046
Description
Reacts with homing-associated cell adhesion molecule (HCAM), CD44, an integral membrane glycoprotein that exists in a variety of isoforms (CD44 variants). CD44 is synthesized as a 37 kDa polypeptide that is converted to a 80-90 kDa form by glycosylation via N- and O-linkages, or to a 180-200 kDa form by the addition of chondroitin sulfate. It is expressed on leukocytes, erythrocytes, epithelial cells and weakly on platelets. Depending on the cell type, CD44 may participate in a variety of functions including cell adhesion, motility and cell activation.
Specifications
| CD44 | |
| Monoclonal | |
| 1.0mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
| Flow Cytometry | |
| L178 | |
| Unconjugated | |
| Mouse | |
| 0.2 mg | |
| Immunology | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.