Learn More
CD43 Mouse anti-Human, Brilliant Violet 421, Clone: 1G10, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 562916
Description
The 1G10 monoclonal antibody specifically binds to CD43, a 95-135 kDa sialoglycoprotein that is highly expressed on most human leucocytes. CD43 is encoded by the SPN gene and is also known as Sialophorin, Galactoglycoprotein (GALGP), Leukosialin, and Leukocyte sialoglycoprotein. The CD43 antigen is expressed on T cells, pre-B cells and activated B cells, NK cells and granulocytes, but is not present on resting peripheral blood B cells, red blood cells, platelets and non-hematopoietic cells. CD43 is enzymatically shed from leucocytesurfaces following activation by various stimuli. CD43 appears to be involved in intercellular interactions that regulate T, B, and NK cell functions. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections with TUF pretreatment.
Specifications
| CD43 | |
| Monoclonal | |
| Brilliant Violet 421 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| 1G10 | |
| SPN; Sialophorin; Galactoglycoprotein/GALGP; GPL115; LEUK; Leukosialin; LSN | |
| Mouse | |
| 100 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.