Learn More
CD42d Mouse anti-Rat, FITC, Clone: RPM.4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 551805
Description
The RPM.4 antibody reacts with CD42d (Platelet glycoprotein V, GPV), an 88-kDa glycoprotein which noncovalently associates with GPIb and GPIX to form a receptor for von Willebrand factor on megakaryocytes and resting platelets. The binding sites for von Willebrand factor and thrombin have been localized to the GPIbα chain of the GPI-b-V-IX complex. GPV is cleaved by thrombin to release a 70 kDa soluble fragment, which contains the epitope recognized by mAb RPM.4. Although the rat CD42d protein's structure is similar to that of human CD42d, the RPM.4 antibody does not cross-react with human platelets.
Specifications
| CD42d | |
| Monoclonal | |
| 0.5mg/mL | |
| Platelet Glycoprotein V | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| RPM.4 | |
| FITC | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Wistar rat platelets | |
| 0.1 mg | |
| Immunology | |
| Rat | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.