Learn More
Invitrogen™ CD42a Monoclonal Antibody (GRP-P), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA181111
Description
For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Human CD42a antibody, clone GRP-P recognizes the platelet GPIX glycoprotein, a 23kDa surface marker expressed by platelets and megakaryocytes. Platelet GPIX is also known as CD42a.The CD42 complex is the major platelet receptor for von Willebrand factor.
Single-chain membrane glycoprotein that forms a non-covalent complex with GPIb. (MW 23 kDa) Reactivity with resting and activated platelets, weakly on monocytes, megakaryocytes and attachment site for the platelet plasma membrane to the submembrane cytoskeleton. GPIb/IX complex, functions as the receptor for ristocetin-induced binding of von Willebrand factor and as the von Willebrand factor-depend adhesion receptor.
Specifications
| CD42a | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 1% BSA and 0.09% sodium azide; pH 7.4 | |
| P14770 | |
| GP9 | |
| Human red blood cells and platelets. | |
| 100 μg | |
| Primary | |
| Human, Mink | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| GRP-P | |
| FITC | |
| GP9 | |
| CD42a; Glycoprotein 9; glycoprotein 9 (platelet); glycoprotein IX (platelet); glycoprotein IX platelet; GP9; GPIX; GP-IX; platelet glycoprotein IX; Platelet GPIX | |
| Mouse | |
| Protein A | |
| RUO | |
| 2815 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.