Learn More
Description
Clone HIP8 reacts with a calcium-dependent complex of CD41/CD61 (GPIIb/IIIa) expressed on platelets and megakaryocytes. CD41/CD61 complex is the receptor of fibrinogen, fibronectin and von Willebrand factor, and mediates platelet adhesion and aggregation. Clone HIP8 also cross-reacts with a major subset of peripheral blood platelets of baboon, and both rhesus and cynomolgus macaque monkeys. The staining pattern of platelets is similar to that observed with peripheral blood platelets from normal human donors.
Specifications
Specifications
| Antigen | CD41a |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIP8 |
| Conjugate | PE |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Immunogen | Purified platelet membrane glycoproteins |
| Purification Method | Affinity Purified |
| Quantity | 50 Tests |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.