missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD41A; Clone: HIP8; BD
Brand: BD Biosciences 563250
Additional Details : Weight : 0.01000kg
Description
Host Species: Mouse
Clone HIP8
Isotype IgG1, κ
Species Reactivity Hu
Immunogen Purified platelet membrane glycoproteins
Flow Cytometry
Specifications
CD41a | |
HIP8 | |
Mouse | |
Affinity Purified | |
RUO | |
Human |
Monoclonal | |
Brilliant Violet 510 | |
Purified platelet membrane glycoproteins | |
50 Tests | |
Primary | |
IgG1 |