missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD40; Clone: 3/23; BD
Brand: BD Biosciences 562847
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone 3/23
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Mouse CD40 Recombinant Protein
Flow Cytometry
Specifications
| CD40 | |
| 3/23 | |
| PE-CF594 | |
| Mouse CD40 Recombinant Protein | |
| 100 μg | |
| Primary | |
| IgG2a |
| Monoclonal | |
| 0.2mg/mL | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction