missing translation for 'onlineSavingsMsg'
Learn More

CD4 Mouse anti-Human, Brilliant Violet 786, Clone: SK3 (also known as Leu3a), BD

Mouse Monoclonal Antibody

£196.31 - £377.08

Specifications

Antigen CD4
Clone SK3 (also known as Leu3a)
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Violet 786
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15843179
View Documents
BD Biosciences
563881
25 Tests
ONLINE EXCLUSIVE PRICE
£199.00
£196.31 / Each
Save £2.69
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15803179
View Documents
BD Biosciences
563877
100 Tests
ONLINE EXCLUSIVE PRICE
£382.00
£377.08 / Each
Save £4.92
1% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The SK3 monoclonal antibody specifically binds to CD4. The CD4 antigen is a 55 kDa type 1 transmembrane glycoprotein that belongs to the immunoglobulin superfamily. It is expressed on T-helper/inducer lymphocytes and monocytes. CD3+CD4+ T cells comprise approximately 28% to 58% of normal peripheral blood lymphocytes. CD4 is also expressed on 80% to 95% of normal thymocytes. The CD4 antigen is present in low density on the cell surface of monocytes and in the cytoplasm of monocytes and macrophages (CD3-CD4+). CD4 serves as a T cell coreceptor for MHC class II antigen recognition and as a receptor for the human immunodeficiency virus (HIV). The SK3 antibody inhibits HIV binding to CD4+ cells.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy7™-like dyes (eg, 780/60-nm filter).

Specifications

Specifications

CD4
Flow Cytometry
Brilliant Violet 786
RUO
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
L3T4; Leu3a; T-cell surface antigen T4/Leu-3 ; W3/25 ; CD4 antigen (p55)
SK3 (also known as Leu3a)
Monoclonal
Mouse
Human
Human Peripheral Blood T Cells
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.