Learn More
CD3G, Mouse, Clone: 2A6, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CD3G.
Brand: Abnova H00000917-M01.100ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is the CD3-gamma polypeptide, which together with CD3-epsilon, -delta and -zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T-cell receptor-CD3 complex. This complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways. The genes encoding the epsilon, gamma and delta polypeptides are located in the same cluster on chromosome 11. Defects in this gene are associated with T cell immunodeficiency. [provided by RefSeq
Sequence: QSIKGNHLVKVYDYQEDGSVLLTCDAEAKNITWFKDGKMIGFLTEDKKKWNLGSNAKDPRGMYQCKGSQNKSKPLQVYYRMCQNCIELNSpecifications
CD3G | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_000073 | |
CD3G | |
CD3G (NP_000064, 23 a.a. ∼ 111 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2A6 | |
Mouse monoclonal antibody raised against a partial recombinant CD3G. | |
CD3G | |
CD3-GAMMA/FLJ17620/FLJ17664/FLJ79544/FLJ94613/MGC138597/T3G | |
Mouse | |
Affinity chromatography | |
RUO | |
917 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |