Learn More
CD3e Mouse anti-Pig, PerCP-Cy5.5, Clone: BB23-8E6-8C8, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561478
Description
The BB23-8E6-8C8 monoclonal antibody specifically binds to the 25-kDa ε chain of the T-cell receptor-associated CD3 complex. It recognizes all CD4+ and most CD8+ peripheral blood T lymphocytes, most thymocytes and phytohemagglutinin-stimulated blasts, and subsets of spleen and Peyer’s patch lymphocytes. BB23-8E6-8C8 is a immunoglobulin isotype switch variant of the BB23-8E6 clone. This isotype-switch variant induces a proliferative response of peripheral blood mononuclear cells. The epitope recognized by BB23-8E6 mAb was designated CD3a by the Second International Swine CD Workshop.
Specifications
| CD3e | |
| Monoclonal | |
| 0.2mg/mL | |
| CD3 epsilon subunit; CD3e; T-cell surface glycoprotein CD3 epsilon chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Pig | |
| IgG2a κ |
| Flow Cytometry | |
| BB23-8E6-8C8 | |
| PerCP-Cyanine5.5 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Pig peripheral blood mononuclear cells | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.