missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ENTPD6 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA548051
Code : NEW
Additional Details : Weight : 0.00010kg
Description
In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CD39 and rhCD39L3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
ENTPD6 Polyclonal antibody specifically detects ENTPD6 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
ENTPD6 | |
Polyclonal | |
Unconjugated | |
Entpd6 | |
2700026H11Rik; CD39 antigen-like 2; CD39L2; CD39-like 2; dJ738P15.3; Ectonucleoside triphosphate diphosphohydrolase 6; ectonucleoside triphosphate diphosphohydrolase 6 (putative); Entpd6; IL-6SAG; IL6ST2; interleukin 6 signal transducer-2; NTPDase 6; NTPDase-6; RP4-738P15.5 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
955 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunoprecipitation, Western Blot | |
0.2 mg/mL | |
PBS with 5% trehalose and No Preservative | |
O75354 | |
Entpd6 | |
CHO-derived recombinant human CD39L2/ENTPD6 Lys61-Ser484 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |