missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD38, BV786; Clone: HIT2; BD
Brand: BD Biosciences 563964
This item is not returnable.
View return policy
Description
Host Species: Mouse
Clone: HIT2
Isotype: IgG1, κ
Species Reactivity: Hu
Flow cytometry
Specifications
| CD38 | |
| HIT2 | |
| Affinity Purified | |
| RUO | |
| Human |
| Monoclonal | |
| Mouse | |
| 100 Tests | |
| Primary |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction