missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVI2B Monoclonal Antibody (MEM-216), PE
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA110213
This item is not returnable.
View return policy
Description
This antibody recognizes an extracellular epitope of CD361 / EVI2B, almost uncharacterized type I transmembrane protein with broad leukocyte expression, mostly in myeloid and B cells.
CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
Specifications
| EVI2B | |
| Monoclonal | |
| PE | |
| EVI2B | |
| CD361; D17S376; ecotropic viral integration site 2B; ecotropic viral integration site 2B protein; Ecotropic viral integration site 2B protein homolog; EVDB; Evi2; Evi-2; EVI2B; EVI-2B; Protein EVI2B; protein EVI2B-like | |
| Mouse | |
| Size-exclusion chromatography | |
| RUO | |
| 2124 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
| Flow Cytometry | |
| MEM-216 | |
| PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
| P34910 | |
| EVI2B | |
| Raji cells. | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction