Learn More
CD36 Mouse anti-Human, FITC, Clone: CB38 (NL07), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555454
Description
The CB38 monoclonal antibody specifically binds to CD36. CD36 is a 88 kDa glycoprotein IV (GPIV), the receptor for extracellular matrix proteins such as collagen and thrombospondin. CD36 is known to mediate the adhesion of Plasmodium falciparum. CD36 antigen is expressed on monocytes, platelets, endothelial cells, and some human tumor cell lines but not on lymphocytes and granulocytes. It is a very early marker of erythroid differentiation. CD36 antibody induces degranulation, release of ATP and serotonin, increase in [Ca2+]ι, and tyrosine phosphorylation of a substrate protein of 130 kDa.
Specifications
| CD36 | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| CB38 (NL07) | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human Platelet Extracts | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.