missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34 Monoclonal Antibody (4H11[APG]), PE, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA119645
Code : LD
Additional Details : Weight : 0.01000kg
Description
CD34 Monoclonal antibody specifically detects CD34 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.Specifications
CD34 | |
Monoclonal | |
0.06 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
P28906 | |
Cd34 | |
Permanent human cell line from a patient with myeloid leukaemia. | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry | |
4H11[APG] | |
PE | |
Cd34 | |
(E3-independent) E2 ubiquitin-conjugating enzyme R1; AU040960; CD34; CD34 antigen; CD34 molecule; CDC34; cell division cycle 34; cell division cycle 34 homolog; cluster designation 34; cluster of differentiation antigen 34; E2 ubiquitin-conjugating enzyme R1; E2-CDC34; hematopoietic progenitor cell antigen CD34; hematopoietic progenitor cell marker CD34; Mucosialin; RP11-328D5.2; sialomucin protein; type 1 transmembrane protein; UBC3; UBCH3; UBE2R1; ubiquitin carrier protein; ubiquitin conjugating enzyme E2 R1; ubiquitin-conjugating enzyme E2 R1; Ubiquitin-conjugating enzyme E2-32 kDa complementing; ubiquitin-conjugating enzyme E2-CDC34; Ubiquitin-protein ligase R1 | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
947 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |