Learn More
CD34 (gp 105-120) Mouse anti-Human, Unlabeled, Clone: MY10, BD
Description
CD34 (Anti–HPCA-1), clone My10, is derived from the hybridization of mouse Sp2/0-Ag14 cells with spleen cells of BALB/c mice immunized with the human cell line KG-1a.
The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.
The CD34 antigen is a single-chain transmembrane glycoprotein, Mr 105 to 120 kilodaltons (kd). The antigen is associated with human hematopoietic progenitor cells and is a differentiation stage–specific leucocyte antigen. Clone My10 and clone 8G12 recognize two distinct CD34 epitopes; at least three epitopes have been identified.
Specifications
Specifications
| Antigen | CD34 (gp 105-120) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | MY10 |
| Concentration | 50μg/mL |
| Conjugate | Unconjugated |
| Formulation | Phosphate buffered saline with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human cell line KG-1a |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.