Learn More
CD335 (NKp46) Mouse anti-Human, Brilliant Violet 786, Clone: 9E2/Nkp46, BD
Mouse Monoclonal Antibody
Brand:  BD Biosciences 563329
Product Code. 15842699
  • ONLINE EXCLUSIVE PRICE
    £310.00
    £296.60 / Each
    Save £13.40
    4% Off
Description
The 9E2/Nkp46 monoclonal antibody specifically binds to CD335. CD335 is also known as the Natural killer cell p46-related protein (NKp46) and the Natural cytotoxicity triggering receptor 1 (NCR1). CD335 is a 46 kDa type I membrane glycoprotein that is expressed on resting and activated NK cells. Its extracel lular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγadaptor proteins. CD335 along with NKp30 and NKp44 are referred to as natural cytotoxicity receptors (NCR). These receptors play very important roles in cells that kill virus-infected target cells, tumor cells and MHC-class I-unprotected cells.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon™ Brilliant Violet™ family of dyes. This dye is a tandem fluorochrome of BD Horizon™ BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon™ BV786 can be excited by the violet laser and detected in a filter used to detect Cy7™-like dyes (eg, 780/60-nm filter).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD335
Monoclonal
Brilliant Violet 786
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human NKp46 Recombinant Protein
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
9E2/Nkp46
NCR1; NK-p46; hNKp46; LY94; Natural cytotoxicity triggering receptor 1
Mouse
Affinity Purified
RUO
Human
IgG1 κ