Learn More
CD335 (NKp46) Mouse anti-Human, Brilliant Violet 510, Clone: 9E2/Nkp46, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 564064
Description
The 9E2/Nkp46 monoclonal antibody specifically binds to CD335. CD335 is also known as the Natural killer cell p46-related protein (NKp46) and the Natural cytotoxicity triggering receptor 1 (NCR1). CD335 is a 46 kDa type I membrane glycoprotein that is expressed on resting and activated NK cells. Its extracel lular region contains two C2-type, Ig-like domains. The transmembrane domain contains a positively charged amino acid (Arg) which could be involved in stabilizing the association with CD3ζ. Its intracellular region does not contain immunoreceptor tyrosine-based activating motifs (ITAM), but it is linked to intracytoplasmic transduction machinery by its association with CD3ζ and FcεRIγadaptor proteins. CD335 along with NKp30 and NKp44 are referred to as natural cytotoxicity receptors (NCR). These receptors play very important roles in cells that kill virus-infected target cells, tumor cells and MHC-class I-unprotected cells.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant Violet™ family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
| CD335 | |
| Monoclonal | |
| Brilliant Violet 510 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Human NKp46 Recombinant Protein | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 9E2/Nkp46 | |
| NCR1; NK-p46; hNKp46; LY94; Natural cytotoxicity triggering receptor 1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.