missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD326; Clone: G8.8; BD
Brand: BD Biosciences 563134
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone G8.8
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line
Flow Cytometry
Specifications
| CD326 | |
| G8.8 | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse |
| Monoclonal | |
| 0.2mg/mL | |
| Glycoconjugates from BALB/c mouse-derived TE-71 medullary thymic epithelial cell line | |
| 50 μg | |
| Primary | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction