Learn More
anti-CD31, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB28364-500ul
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Human
Immunogen: Synthetic peptide corresponding to C terminus of mouse CD31
Applications:
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| CD31 | |
| 0.2mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cancer and Tumor Biology, Cardiovascular, Signal Transduction, Stem Cell Biology | |
| Human, Mouse |
| Polyclonal | |
| Unconjugated | |
| Synthetic peptide corresponding to C terminus of mouse CD31 | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.