missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD31; Clone: MEC 13.3; BD
Brand: BD Biosciences 562861
This item is not returnable.
View return policy
Description
Host Species: Rat
Clone MEC 13.3
Isotype IgG2a, κ
Species Reactivity Ms
Immunogen 129/Sv mouse-derived endothelioma cell line tEnd.1
Flow Cytometry
Specifications
| CD31 | |
| MEC 13.3 | |
| PerCP-Cyanine5.5 | |
| 129/Sv mouse-derived endothelioma cell line tEnd.1 | |
| 100 μg | |
| Primary | |
| IgG2a |
| Monoclonal | |
| 0.2mg/mL | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction